6-Chloro-4-(trifluoromethyl)pyridin-2-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-hydroxy-4-(trifluoromethyl)pyridine

058930-500MG 058930-1G
101916-370EA 167.69 USD
101916-370 101916-372
6-Chloro-4-(trifluoromethyl)pyridin-2-ol ≥95%
6-Chloro-4-(trifluoromethyl)pyridin-2-ol

Hazard: Irritant

Formula: C₆H₃ClF₃NO
MW: 197.55 g/mol
MDL Number: MFCD13190002
CAS Number: 34486-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR