[(6-Chloro-4-trifluoromethylpyridin-2-yl)amino]acetic acid ethyl ester ≥95%

Supplier: Matrix Scientific

058929-500MG 058929-1G
101916-366EA 553.85 USD
101916-366 101916-368
[(6-Chloro-4-trifluoromethylpyridin-2-yl)amino]acetic acid ethyl ester ≥95%
[(6-Chloro-4-trifluoromethylpyridin-2-yl)amino]acetic acid ethyl ester

Hazard: Irritant

Formula: C₁₀H₁₀ClF₃N₂O₂
MDL Number: MFCD15142771
CAS Number: 1208081-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR