2-Chloro-5-trifluoromethyl[1,3,4]thiadiazole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-trifluoromethyl-[1,3,4]-thiadiazole

058924-1G 058924-5G
101916-348EA 66.15 USD
101916-348 101916-350
2-Chloro-5-trifluoromethyl[1,3,4]thiadiazole ≥95%
2-Chloro-5-trifluoromethyl[1,3,4]thiadiazole

Hazard: Irritant

Formula: C₃ClF₃N₂S
MW: 188.56 g/mol
MDL Number: MFCD04972766
CAS Number: 53645-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR