2-(3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulfanyl)propionic acid ≥95%

Supplier: Matrix Scientific

058923-1G 058923-5G
101916-344EA 573.85 USD
101916-344 101916-346
2-(3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulfanyl)propionic acid ≥95%
2-(3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulfanyl)propionic acid

Hazard: Irritant

Formula: C₉H₇ClF₃NO₂S
MDL Number: MFCD09949812
CAS Number: 1016845-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR