2-(3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulfanyl)ethanol ≥95%

Supplier: Matrix Scientific

058922-1G 058922-5G 058922-10G
101916-338EA 278.46 USD
101916-338 101916-340 101916-342
2-(3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulfanyl)ethanol ≥95%
2-(3-Chloro-5-(trifluoromethyl)pyridin-2-ylsulfanyl)ethanol

Hazard: Irritant

Formula: C₈H₇ClF₃NOS
MDL Number: MFCD10568271
CAS Number: 188591-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR