2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)ethanol ≥95%

Supplier: Matrix Scientific

058921-1G 058921-5G 058921-10G
101916-332EA 378.46 USD
101916-332 101916-334 101916-336
2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)ethanol ≥95%
2-(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)ethanol

Hazard: Irritant

Formula: C₈H₇ClF₃NO₂
MDL Number: MFCD10568241
CAS Number: 95355-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR