(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)acetic acid

Supplier: Matrix Scientific

058920-1G 058920-5G
101916-328EA 400 USD
101916-328 101916-330
(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)acetic acid
(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)acetic acid

Hazard: Irritant

Formula: C₈H₅ClF₃NO₃
MDL Number: MFCD10568276
CAS Number: 95355-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR