2-Chloro-4-(3-trifluoromethylphenyl)[1,3,5]triazine ≥95%

Supplier: Matrix Scientific

058918-1G 058918-5G
101916-320EA 546.15 USD
101916-320 101916-322
2-Chloro-4-(3-trifluoromethylphenyl)[1,3,5]triazine ≥95%
2-Chloro-4-(3-trifluoromethylphenyl)[1,3,5]triazine

Hazard: Irritant

Formula: C₁₀H₅ClF₃N₃
MW: 259.62 g/mol
MDL Number: MFCD11052414
CAS Number: 1053658-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR