2-(6-Chloro-4-(trifluoromethyl)pyridin-2-ylsulfanyl)propionic acid ≥95%

Supplier: Matrix Scientific

058917-1G 058917-5G
101916-316EA 669.23 USD
101916-316 101916-318
2-(6-Chloro-4-(trifluoromethyl)pyridin-2-ylsulfanyl)propionic acid ≥95%
2-(6-Chloro-4-(trifluoromethyl)pyridin-2-ylsulfanyl)propionic acid

Hazard: Irritant

Formula: C₉H₇ClF₃NO₂S
MDL Number: MFCD11052386
CAS Number: 1053656-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR