2-(6-Chloro-4-(trifluoromethyl)pyridin-2-ylsulfanyl)acetic acid ≥95%

Supplier: Matrix Scientific

058916-1G 058916-5G
101916-312EA 426.15 USD
101916-312 101916-314
2-(6-Chloro-4-(trifluoromethyl)pyridin-2-ylsulfanyl)acetic acid ≥95%
2-(6-Chloro-4-(trifluoromethyl)pyridin-2-ylsulfanyl)acetic acid

Hazard: Irritant

Formula: C₈H₅ClF₃NO₂S
MDL Number: MFCD11052387
CAS Number: 1053656-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR