N-(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)-N-methylhydrazine ≥95%

Supplier: Matrix Scientific

058915-1G 058915-5G
101916-308EA 553.85 USD
101916-308 101916-310
N-(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)-N-methylhydrazine ≥95%
N-(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)-N-methylhydrazine

Hazard: Irritant

Formula: C₇H₇ClF₃N₃
MDL Number: MFCD11052356
CAS Number: 1053659-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR