4-(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)morpholine ≥95%

Supplier: Matrix Scientific

058914-1G 058914-5G
101916-304EA 393.85 USD
101916-304 101916-306
4-(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)morpholine ≥95%
4-(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)morpholine

Hazard: Irritant

Formula: C₁₀H₁₀ClF₃N₂O
MDL Number: MFCD09991798
CAS Number: 1053659-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR