(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)hydrazine ≥95%

Supplier: Matrix Scientific

058913-1G 058913-5G
101916-300EA 553.85 USD
101916-300 101916-302
(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)hydrazine ≥95%
(6-Chloro-4-(trifluoromethyl)pyridin-2-yl)hydrazine

Hazard: Irritant

Formula: C₆H₅ClF₃N₃
MDL Number: MFCD10568286
CAS Number: 129015-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR