1-(4-Chloropyridin-2-yl)ethanone ≥95%

Supplier: Matrix Scientific

058905-1G 058905-5G
101916-270EA 415.38 USD
101916-270 101916-272
1-(4-Chloropyridin-2-yl)ethanone ≥95%
1-(4-Chloropyridin-2-yl)ethanone

Hazard: Irritant

Formula: C₇H₆ClNO
MDL Number: MFCD10697662
CAS Number: 60159-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR