(6-Chloro-pyridin-3-yl)carbamic acid tert-butyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl (6-chloropyridin-3-yl)carbamate

058904-1G 058904-5G
101916-266EA 116.92 USD
101916-266 101916-268
(6-Chloro-pyridin-3-yl)carbamic acid tert-butyl ester ≥95%
(6-Chloro-pyridin-3-yl)carbamic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MW: 228.68 g/mol
MDL Number: MFCD08062421
CAS Number: 171178-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR