2-Chloropyridine-3-sulfonic acid amide ≥95%

Supplier: Matrix Scientific

058902-1G 058902-5G
101916-262EA 316.92 USD
101916-262 101916-264
2-Chloropyridine-3-sulfonic acid amide ≥95%
2-Chloropyridine-3-sulfonic acid amide

Hazard: Irritant

Formula: C₅H₅ClN₂O₂S
MDL Number: MFCD08752623
CAS Number: 38025-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR