4-Chloropyridine-N-oxide ≥95%

Supplier: Matrix Scientific

058901-1G 058901-5G 058901-10G
101916-256EA 175.38 USD
101916-256 101916-258 101916-260
4-Chloropyridine-N-oxide ≥95%
4-Chloropyridine N-oxide

Hazard: Irritant

Formula: C₅H₄ClNO
MDL Number: MFCD00047425
CAS Number: 1121-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR