4-Chloropyridine-2-carbonitrile ≥95%

Supplier: Matrix Scientific

058900-1G 058900-5G
101916-252EA 15.38 USD
101916-252 101916-254
4-Chloropyridine-2-carbonitrile ≥95%
4-Chloropyridine-2-carbonitrile

Hazard: Irritant

Formula: C₆H₃ClN₂
MW: 138.5500 g/mol
MDL Number: MFCD06009833
CAS Number: 19235-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR