4-(3-Chloropropyl)morpholine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Morpholinopropyl Chloride

058899-5G 058899-25G
101916-248EA 15.38 USD
101916-248 102551-550
4-(3-Chloropropyl)morpholine ≥95%
4-(3-Chloropropyl)morpholine

Hazard: Irritant

Formula: C₇H₁₄ClNO
MW: 163.65 g/mol
MDL Number: MFCD00039714
CAS Number: 7357-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR