N'-[2-(4-Chlorophenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl ≥95%

Supplier: Matrix Scientific

058898-500MG 058898-1G
101916-242EA 284.62 USD
101916-242 101916-244
N'-[2-(4-Chlorophenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl ≥95%
N'-[2-(4-Chlorophenyl)-1-(2,2,2-trifluoroethylamino)ethylidene]hydrazinecarboxylic acid-tert-butyl

Hazard: Irritant

Formula: C₁₅H₁₉ClF₃N₃O₂
MW: 365.79 g/mol
MDL Number: MFCD10568263
CAS Number: 1053658-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR