4-Chloro-2-phenylthiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

058897-1G 058897-5G
101916-238EA 495.38 USD
101916-238 101916-240
4-Chloro-2-phenylthiazole-5-carbaldehyde ≥95%
4-Chloro-2-phenylthiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₆ClNOS
MDL Number: MFCD01109238
CAS Number: 108263-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR