5-Chloro-3-phenyl[1,2,4]thiadiazole ≥95%

Supplier: Matrix Scientific

058896-1G 058896-5G
101916-232EA 193.85 USD
101916-232 101916-234
5-Chloro-3-phenyl[1,2,4]thiadiazole ≥95%
5-Chloro-3-phenyl[1,2,4]thiadiazole

Hazard: Irritant

Formula: C₈H₅ClN₂S
MDL Number: MFCD00096450
CAS Number: 24255-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR