2-(4-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: α-(4-Morpholino)-4-chlorophenylacetonitrile hydrochloride

058895-1G 058895-5G 058895-10G
101916-226EA 204.62 USD
101916-226 101916-228 101916-230
2-(4-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile hydrochloride ≥95%
2-(4-Chlorophenyl)-2-(morpholin-4-yl)acetonitrile hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O·ClH
MW: 273.158 g/mol
MDL Number: MFCD11052434
CAS Number: 1330171-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR