N'-[2-(4-Chlorophenyl)-1-cyclobutylaminoethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058888-500MG 058888-1G
101916-202EA 284.62 USD
101916-202 101916-204
N'-[2-(4-Chlorophenyl)-1-cyclobutylaminoethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[2-(4-Chlorophenyl)-1-cyclobutylaminoethylidene]hydrazinecarboxylic acid-tert-butyl ester

Hazard: Irritant

Formula: C₁₇H₂₄ClN₃O₂
MDL Number: MFCD10568251
CAS Number: 1053657-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR