2-(4-Chlorophenyl)amino-4-phenylpyrimidine ≥95%

Supplier: Matrix Scientific

058886-1G 058886-5G
101916-196EA 256.92 USD
101916-196 101916-198
2-(4-Chlorophenyl)amino-4-phenylpyrimidine ≥95%
2-(4-Chlorophenyl)amino-4-phenylpyrimidine

Hazard: Irritant

Formula: C₁₆H₁₂ClN₃
MDL Number: MFCD10568239
CAS Number: 1053657-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR