4'-(6-Chloro-5-nitro-4-(trifluoromethyl)pyridin-2-yl)morpholine ≥95%

Supplier: Matrix Scientific

058884-1G 058884-5G
101916-188EA 669.23 USD
101916-188 101916-190
4'-(6-Chloro-5-nitro-4-(trifluoromethyl)pyridin-2-yl)morpholine ≥95%
4'-(6-Chloro-5-nitro-4-(trifluoromethyl)pyridin-2-yl)morpholine

Hazard: Irritant

Formula: C₁₀H₉ClF₃N₃O₃
MW: 311.65 g/mol
MDL Number: MFCD11052355
CAS Number: 1053659-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR