6-Chloronicotinamidine hydrochloride ≥95%

Supplier: Matrix Scientific

058882-1G 058882-5G
101916-180EA 436.92 USD
101916-180 101916-182
6-Chloronicotinamidine hydrochloride ≥95%
6-Chloronicotinamidine hydrochloride

Hazard: Irritant

Formula: C₆H₇Cl₂N₃
MDL Number: MFCD11052371
CAS Number: 201937-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR