6-Chloro-N-hydroxynicotinamidine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-N-hydroxynicotinimidamide

058881-1G 058881-5G 058881-10G
101916-174EA 263.08 USD
101916-174 101916-176 101916-178
6-Chloro-N-hydroxynicotinamidine ≥95%
6-Chloro-N-hydroxynicotinamidine

Hazard: Irritant

Formula: C₆H₆ClN₃O
MW: 171.59 g/mol
MDL Number: MFCD11052369
CAS Number: 468068-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR