2-Chloro-5-methylpyridine-3-sulfonyl chloride ≥95%

Supplier: Matrix Scientific

058878-500MG 058878-1G
101916-162EA 363.08 USD
101916-162 101916-164
2-Chloro-5-methylpyridine-3-sulfonyl chloride ≥95%
2-Chloro-5-methylpyridine-3-sulfonyl chloride

Hazard: Irritant

Formula: C₆H₅Cl₂NO₂S
MDL Number: MFCD15142818
CAS Number: 1208081-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR