2-Chloromethyl-6-nitro-1H-benzoimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Chloromethyl)-6-nitro-1H-benzo[d]imidazole

058875-1G 058875-5G 058875-10G
101916-146EA 378.46 USD
101916-146 101916-148 101916-150
2-Chloromethyl-6-nitro-1H-benzoimidazole ≥95%
2-Chloromethyl-6-nitro-1H-benzoimidazole

Hazard: Irritant

Formula: C₈H₆ClN₃O₂
MW: 211.61 g/mol
MDL Number: MFCD01785201
CAS Number: 14625-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR