2-Chloro-3-cyano-4-picoline ≥95%

Supplier: Matrix Scientific

058874-1G 058874-250MG
101916-144EA 153.85 USD
101916-144 102833-270
2-Chloro-3-cyano-4-picoline ≥95%
2-Chloro-3-cyano-4-picoline

Hazard: Irritant

Formula: C₇H₅ClN₂
MDL Number: MFCD07082888
CAS Number: 65169-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR