3-Chloromethyl-5-methyl[1,2,4]oxadiazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Chloromethyl)-5-methyl-1,2,4-oxadiazole

058870-1G 058870-5G
101916-128EA 115.38 USD
101916-128 101916-130
3-Chloromethyl-5-methyl[1,2,4]oxadiazole ≥95%
3-Chloromethyl-5-methyl[1,2,4]oxadiazole

Hazard: Irritant

Formula: C₄H₅ClN₂O
MW: 132.55 g/mol
MDL Number: MFCD01809083
CAS Number: 1192-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR