5-Chloromethyl-2-methyl-2H-pyrazole-3-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

058869-500MG 058869-1G 058869-5G
101916-122EA 418.46 USD
101916-122 101916-124 101916-126
5-Chloromethyl-2-methyl-2H-pyrazole-3-carboxylic acid methyl ester ≥95%
5-Chloromethyl-2-methyl-2H-pyrazole-3-carboxylic acid methyl ester

Hazard: Irritant

Formula: C₇H₉ClN₂O₂
MDL Number: MFCD11845756
CAS Number: 1208081-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR