5-(Chloromethyl)-3-(4-methoxyphenyl)-1,2,4-oxadiazole ≥95%

Supplier: Matrix Scientific

058868-250MG 058868-1G
101916-118EA 175.38 USD
101916-118 101916-120
5-(Chloromethyl)-3-(4-methoxyphenyl)-1,2,4-oxadiazole ≥95%
5-(Chloromethyl)-3-(4-methoxyphenyl)-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₁₀H₉ClN₂O₂
CAS Number: 57238-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR