4-Chloro-5-methyl-isoxazol-3-ylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-4-chloro-5-methylisoxazole

058867-1G 058867-5G
101916-114EA 258.46 USD
101916-114 101916-116
4-Chloro-5-methyl-isoxazol-3-ylamine ≥95%
4-Chloro-5-methyl-isoxazol-3-ylamine

Hazard: Irritant

Formula: C₄H₅ClN₂O
MW: 132.55 g/mol
MDL Number: MFCD11227222
CAS Number: 5819-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR