4-Chloro-3-methylisothiazol-5-ylamine 95

Supplier: Matrix Scientific

058866-500MG 058866-1G 058866-5G
101916-108EA 466.15 USD
101916-108 101916-110 101916-112
4-Chloro-3-methylisothiazol-5-ylamine 95
4-Chloro-3-methylisothiazol-5-ylamine

Hazard: Irritant

Formula: C₄H₅ClN₂S
MW: 148.61 g/mol
MDL Number: MFCD09909657
CAS Number: 96841-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR