tert-Butyl-3-(chloromethyl)benzoate ≥90%

Supplier: Matrix Scientific

Synonyms: 3-Chloromethyl-benzoic acid tert-butyl ester

058860-1G 058860-5G
101916-080EA 172.31 USD
101916-080 101916-082
tert-Butyl-3-(chloromethyl)benzoate ≥90%
tert-Butyl-3-(chloromethyl)benzoate

Hazard: Irritant

Formula: C₁₂H₁₅ClO₂
MDL Number: MFCD02180879
CAS Number: 220510-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR