2-Acetyl-5-chloro-3-methylbenzo[b]thiophene ≥95%

Supplier: Matrix Scientific

058859-1G 058859-5G
101916-076EA 146.15 USD
101916-076 101916-078
2-Acetyl-5-chloro-3-methylbenzo[b]thiophene ≥95%
2-Acetyl-5-chloro-3-methylbenzo[b]thiophene

Hazard: Irritant

Formula: C₁₁H₉ClOS
CAS Number: 51527-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR