2-Chloro-6-methoxypyridine-3-sulfonyl chloride ≥95%
Supplier: Matrix Scientific
058856-500MG
058856-1G
101916-064EA
518.46
USD
101916-064
101916-066
2-Chloro-6-methoxypyridine-3-sulfonyl chloride ≥95%
2-Chloro-6-methoxypyridine-3-sulfonyl chloride
Hazard: Corrosive
Formula:
C₆H₅Cl₂NO₃S Melting Pt: 94…95 °C |
MDL Number:
MFCD15142789 CAS Number: 1208081-26-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...