2-Chloroisonicotinoyl chloride ≥95%

Supplier: Matrix Scientific

058855-1G 058855-5G 058855-10G
101916-058EA 175.38 USD
101916-058 101916-060 101916-062
2-Chloroisonicotinoyl chloride ≥95%
2-Chloroisonicotinoyl chloride

Hazard: Corrosive/Stored under Argon

Formula: C₆H₃Cl₂NO
Boiling Pt: 144-148°C/10mmHg
Density: 1.418
MDL Number: MFCD00204232
CAS Number: 65287-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR