2-Chloroisonicotinamidine hydrochloride ≥95%

Supplier: Matrix Scientific

058854-1G 058854-5G
101916-054EA 135.38 USD
101916-054 101916-056
2-Chloroisonicotinamidine hydrochloride ≥95%
2-Chloroisonicotinamidine hydrochloride

Hazard: Irritant

Formula: C₆H₇Cl₂N₃
MW: 192.05 g/mol
MDL Number: MFCD11052370
CAS Number: 82019-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR