3-Chloro-5-iodopyridin-2-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-5-iodo-pyridin-2-ol

058853-1G 058853-5G
101916-050EA 532.31 USD
101916-050 101916-052
3-Chloro-5-iodopyridin-2-ol ≥95%
3-Chloro-5-iodopyridin-2-ol

Hazard: Irritant

Formula: C₅H₃ClINO
MW: 255.44 g/mol
MDL Number: MFCD11227187
CAS Number: 97966-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR