2-Chloro-6-hydroxynicotinic acid ≥95%

Supplier: Matrix Scientific

058852-1G 058852-5G 058852-25G
101916-046EA 138.46 USD
101916-046 101916-048 102551-548
2-Chloro-6-hydroxynicotinic acid ≥95%
2-Chloro-6-hydroxynicotinic acid

Hazard: Irritant

Formula: C₆H₄ClNO₃
MW: 173.5500 g/mol
Melting Pt: 300-302°C
MDL Number: MFCD09757508
CAS Number: 38025-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR