5-Chloro-4,6-dimethylpyrimidin-2-ol ≥95%

Supplier: Matrix Scientific

058846-500MG 058846-1G 058846-5G
101916-026EA 453.85 USD
101916-026 101916-028 101916-030
5-Chloro-4,6-dimethylpyrimidin-2-ol ≥95%
5-Chloro-4,6-dimethylpyrimidin-2-ol

Hazard: Irritant

Formula: C₆H₇ClN₂O
MW: 158.59 g/mol
Melting Pt: 241…245 °C
MDL Number: MFCD12131131
CAS Number: 19573-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR