6-Chloro-2,5-dimethylpyrimidine-4-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

058845-1G 058845-5G
101916-022EA 495.38 USD
101916-022 101916-024
6-Chloro-2,5-dimethylpyrimidine-4-carboxylic acid ethyl ester ≥95%
6-Chloro-2,5-dimethylpyrimidine-4-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂
MW: 214.65 g/mol
MDL Number: MFCD02180838
CAS Number: 76498-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR