2-Chloro-4,6-dimethylnicotinamide ≥95%

Supplier: Matrix Scientific

058844-1G 058844-10G
101916-018EA 264.62 USD
101916-018 101916-020
2-Chloro-4,6-dimethylnicotinamide ≥95%
2-Chloro-4,6-dimethylnicotinamide

Hazard: Irritant

Formula: C₈H₉ClN₂O
MDL Number: MFCD00067869
CAS Number: 140413-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR