5-Chloro-2-cyclopropyl-6-methylpyrimidin-4-ol ≥95%

Supplier: Matrix Scientific

058843-1G 058843-5G 058843-10G
101916-012EA 256.92 USD
101916-012 101916-014 101916-016
5-Chloro-2-cyclopropyl-6-methylpyrimidin-4-ol ≥95%
5-Chloro-2-cyclopropyl-6-methylpyrimidin-4-ol

Hazard: Irritant

Formula: C₈H₉ClN₂O
MDL Number: MFCD00114339
CAS Number: 287927-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR