5-Chloro-3-chloromethyl[1,2,4]thiadiazole ≥95%

Supplier: Matrix Scientific

058840-1G 058840-5G
101916-000EA 187.69 USD
101916-000 101916-002
5-Chloro-3-chloromethyl[1,2,4]thiadiazole ≥95%
5-Chloro-3-chloromethyl[1,2,4]thiadiazole

Hazard: Irritant

Formula: C₃H₂Cl₂N₂S
MDL Number: MFCD00096470
CAS Number: 74461-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR