2-Chloro-4-chloromethylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-(chloromethyl)pyridine

058839-1G 058839-5G
101915-996EA 95.38 USD
101915-996 101915-998
2-Chloro-4-chloromethylpyridine ≥95%
2-Chloro-4-chloromethylpyridine

Hazard: Irritant

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
MDL Number: MFCD09991655
CAS Number: 101990-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR