3-Chloro-2-(2-chloroethoxy)-5-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

058838-1G 058838-5G
101915-990EA 436.92 USD
101915-990 101915-992
3-Chloro-2-(2-chloroethoxy)-5-(trifluoromethyl)pyridine ≥95%
3-Chloro-2-(2-chloroethoxy)-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₈H₆Cl₂F₃NO
MW: 260.04 g/mol
MDL Number: MFCD10568240
CAS Number: 101456-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR